Learn More
Enzo Life Sciences Pepstatin A (synthetic) (25 mg) CAS: 26305-03-3
SDP
Supplier:  Enzo Life Sciences ALX260085M025
Encompass_Preferred
Protease inhibitor. Alternative Name: Iva-Val-Val-Sta-Ala-Sta, i-Valeryl-L-Val-L-Val-AHMHA-L-Ala-AHMHA. Formula: C34H63N5O9. MW: 685.9. CAS: 26305-03-3. MI: 14: 7147. Purity: ≥98% (HPLC). Sequence: i-Valeryl-L-Val-L-Val-AHMHA-L-Ala-AHMHA. (AHMHA = (3S,4S)-4-Amino-3-hydroxy-6-methyl-heptanoic acid). Appearance: White to off-white solid. Solubility: Soluble in DMSO (10mg/ml), methanol (1mg/ml) or acetic acid. Long Term Storage: -20°C. Use/Stability: Stable for at least 2 years after receipt when stored at -20°C. Handling: Protect from light.
Catalog No. NC1460915