Learn More
Takara Bio PEPTIDE COATING KIT 96 RXNS
SDP
Supplier:  Takara Bio MK100
Encompass
Peptide Coating Kit; Designed for easy and efficient coating of peptides on plates for subsequent use in ELISA; Includes five 96-well strip plates, 5mL coupling reagent, 50mL reaction buffer, and 2 x 50mL blocking solution
Catalog No. 50-443-931