Learn More
AnaSpec Peptide Mass Spec Standards, (1 kit)
Supplier:  AnaSpec AS60882
Encompass
Peptide Mass Spec Standards, (1 kit) This kit consists of two calibration mixtures for calibrating mass scale in MALDI-TOF or ESI mass spectrometry. Calibration Mixture 1 ranges from 800 to 1800 daltons, and Calibration Mixture 2 from 1800 to 3800 daltons.
Catalog No. NC9931266