Learn More
Bachem Peptide YY (3-36) (human) Trifluoroacetate
SDP
Supplier:  Bachem 4018880.0500
Encompass_Preferred
Peptide YY (3-36) (human) Trifluoroacetate, H-Ile-Lys-Pro-Glu-Ala-Pro-Gly-Glu-Asp-Ala-Ser-Pro-Glu-Glu-Leu-Asn-Arg-Tyr-Tyr-Ala-Ser-Leu-Arg-His-Tyr-Leu-Asn-Leu-Val-Thr-Arg-Gln-Arg-Tyr-NH2, 123583-37-9, C180H279N53O54, Mr 4049.52, 0.5mg. PYY (3-36) (human). PYY (3-36), a Y2 receptor agonist, is released from the body's gastrointestinal tract in proportion to caloric intake. It has been shown that peripheral injection of PYY (3-36) in rats inhibited food intake and reduced weight gain. In addition, infusion of PYY (3-36) in humans significantly decreased appetite and reduced food intake by 33% over 24h, which suggests that PYY (3-36) has a role in 'longer term' regulation of food intake. Thus, the PYY (3-36) may represent a lead compound for the development of drugs for the treatment of obesity.
Catalog No. 50-260-197