Learn More
CPC Scientific H-Ile-Lys-Pro-Glu-Ala-Pro-Gly-Glu-Asp-Ala-Ser-Pro-Glu-Glu-Leu-Asn-Arg-Tyr-Tyr-Ala-Ser-Leu-Arg-His-Tyr-Leu-Asn-Leu-Val-Thr-Arg-Gln-Arg-Tyr-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PEPY005B
Encompass
Y2 receptor agonist, released from the body's gastrointestinal tract in proportion to caloric intake, that significantly decreases appetite and reduces food intake, and thus may act as a drug for the treatment of obesity. ONE-LETTER SEQUENCE: IKPEAPGEDASPEELNRYYASLRHYLNLVTRQRY-NH2MOLECULAR FORMULA: C180H279N53O54MOLECULAR WEIGHT:4049.52STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [123583-37-9], SYNONYMS: PYY (3-36) (human)Peptide YY (3-36) (human)RESEARCH AREA: GastrointestinalREFERENCES: D. Grandt et al., Regulatory Peptides, 40, 161(1992)
Catalog No. 50-211-0149