Learn More
CPC Scientific H-Tyr-Pro-Ala-Lys-Pro-Glu-Ala-Pro-Gly-Glu-Asp-Ala-Ser-Pro-Glu-Glu-Leu-Ser-Arg-Tyr-Tyr-Ala-Ser-Leu-Arg-His-Tyr-Leu-Asn-Leu-Val-Thr-Arg-Gln-Arg-Tyr-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PEPY004B
Encompass
ONE-LETTER SEQUENCE: YPAKPEAPGEDASPEELSRYYASLRHYLNLVTRQRY-NH2MOLECULAR FORMULA: C190H288N54O57MOLECULAR WEIGHT:4240.71STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [81858-94-8], SYNONYMS: PYY (canine, mouse, porcine, rat), Peptide YY (canine, mouse, porcine, rat)RESEARCH AREA: GastrointestinalREFERENCES: K. Tatemoto et al., Nature, 285, 417 (1980); R. Corder et al., Regulatory Peptides, 21, 253 (1988); K. Tatemoto, PNAS, 79, 2514 (1982)
Catalog No. 50-211-0147