Learn More
Medchemexpress LLC Peptide ready HLA-A*02:01 & B2M monomer protein, human (HEK293, His-Avi) | 50UG
SDP
Supplier:  Medchemexpress LLC HYP70284050UG
Encompass_Preferred
Peptide Ready HLA-A*02:01&B2M Monomer Protein is a recombinant human MHC class I monomer expressed in HEK293 cells, engineered with C-terminal His and Avi tags for downstream conjugation. It is supplied for research use and designed to be peptide-loadable and compatible with tetramerization workflows, provided as a 50 μg vial for laboratory applications.
  • Recombinant human HLA-A*02:01 complexed with beta-2 microglobulin.
  • Expressed in HEK293 mammalian cells for native glycosylation.
  • C-terminal His and Avi tags for purification and biotinylation.
  • Supplied as a monomer ready for peptide loading.
  • Compatible with tetramerization and immunological assays.
  • Provided in a small pack size suitable for research workflows.
  • Approximate molecular weight 43 kDa under reducing conditions.
Catalog No. 50-001-74270