Learn More
Waters Corp PeptideWorks Tryptic Start-Up & Column Kit, 30 min. Digestion, 96 Sample Throughput, MS Grade, CSH C18 Column
Supplier:  Waters Corp 176005310
Encompass_Preferred
This kit offers an automatable, high-efficiency, and reproducible method for peptide mapping, delivering results in under 2.5 hr., four times faster than average home-brew methods. It supports manual or automated protein digestion, featuring MS Grade RapiZyme Trypsin for complete 30 min. digestions without contaminating autolysis peaks. The kit includes all components for protein denaturation, reduction, alkylation, desalting, and digestion, supporting 4 to 96 samples.
  • Complete protein digestions in 30 min.
  • Processes 4 to 96 samples.
  • MS Grade RapiZyme Trypsin ensures clean digestions.
  • Includes a peptide CSH C18 column for LC separation.
  • All reagents for denaturation, reduction, alkylation, desalting, and digestion included.
  • Automated or manual workflow completes in under 2.5 hr.
Catalog No. 50-239-2841