Learn More
TOKU-E Blasticidin S Hydrochloride, 3513-03-9, 1 g
Supplier:  TOKU-E B0011G
Encompass_Preferred
peptidyl nucleoside antibiotic, C17H26N8O5 HCl, selection antibiotic, Inhibits protein synthesis. Useful in cell culture, microbiology.
Catalog No. 50-255-1832