Learn More
Medchemexpress LLC Peptidyl-prolyl cis-trans isomerase A/CYPA protein, human | >95.0% | 100UG

Supplier: Medchemexpress LLC HYP70047100UG
Recombinant human peptidyl-prolyl cis-trans isomerase A (Cyclophilin A) supplied as a 0.2 μm-filtered protein solution for research use. The protein is tag-free, characterized for purity and molecular weight, and formulated for stability in storage and assays.
- Purity >95.0% as determined by reducing SDS-PAGE.
- Supplied in PBS with 10% glycerol, pH 7.4, 0.2 μm filtered.
- Tag free protein expressed in Escherichia coli.
- Molecular weight 16-18 kDa.
- Store at -80°C for long-term stability; stable at -20°C for 3 months after opening.
- Avoid repeated freeze-thaw cycles; aliquot for extended storage.
- Sequence available for verification and analysis.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.