Learn More
Medchemexpress LLC Peptidyl-prolyl cis-trans isomerase A (Cyclophilin A) | >95.0% | 250UG
SDP
Supplier:  Medchemexpress LLC HYP70047A250UG
Encompass_Preferred
Peptidyl-prolyl cis-trans isomerase A (cyclophilin A) is a recombinant, tag-free human protein provided as a purified solution for research applications. It catalyzes cis-trans isomerization of proline residues and is suitable for biochemical assays and protein interaction studies.
Catalog No. 50-001-76818