Learn More
Medchemexpress LLC Peptidyl-prolyl cis-trans isomerase A/CYPA protein, human | >95.0% | 500UG
SDP
Supplier:  Medchemexpress LLC HYP70047500UG
Encompass_Preferred
Peptidyl-prolyl cis-trans isomerase A (Cyclophilin A) is supplied as a recombinant human protein solution produced in E. coli and formulated for research use. The protein is supplied tag-free, quality-controlled, and intended for biochemical assays, binding studies, and functional characterization of PPIase activity.
Catalog No. 50-001-74931