Learn More
Alkali Scientific Peptone, 1 Gram Capsules, 1 Kg
SDP
Supplier:  Alkali Scientific EPT001CP
Encompass_Preferred
Peptone, available in 1g capsules (1kg total per bottle), is a high-quality protein source commonly used in microbiological and biochemical media. This product provides essential amino acids, peptides, and growth factors required for the cultivation of a wide range of microorganisms. The 1-gram capsules allow for precise and convenient measurement of peptone, ensuring consistent media preparation without the need for bulky containers. It is widely used in media for bacterial growth, fermentation studies, and microbiological quality control. This product is ideal for enhancing the nutritional content of culture media for both research and industrial microbiological applications.
Catalog No. 50-006-59776