Learn More
Research Products International Corp Peptone, Powder, 5 Kilograms
Supplier: Research Products International Corp P202405000.0
Mixture of peptides and free amino acids from pancreatic hydrolysis of animal tissue. Low NaCl content suitable for tissue culture.
Total Nitrogen (TN): 15.0-16.0%
Amino Nitrogen (AN): 3.0-3.5%
AN/TN Ratio: 0.21-0.23