Learn More
Medchemexpress LLC Peptone from soya | 91079-46-8 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW133891100G
Encompass_Preferred
Peptone from soya is a soybean-derived enzymatic digest used as a nutrient supplement for microbiological and cell culture media. It supplies amino acids, peptides, and other growth factors that support microbial proliferation and influence cellular responses. The product is supplied as a light yellow to light brown powder with recommended storage at room temperature.
  • Provides amino acids, peptides, and nitrogen sources for culture media.
  • Supports microbial growth and cell-based assays.
  • Water-soluble; may require sonication for complete dissolution.
  • Offered in multiple pack sizes, including 100 G.
  • Store at room temperature; stable under recommended conditions.
Catalog No. 50-002-83509