Learn More
ATCC Peptostreptococcus anaerobius; Strain VPI 4330
Supplier:  ATCC 27337
Encompass
Peptostreptococcus anaerobius strain VPI 4330 has media testing and quality control applications. This whole-genome sequenced bacterial type strain is propagated anaerobically and requires proline for glucose utilization.
Catalog No. 50-238-4621