Learn More
Cygnus Technologies Cygnus Technologies PER.C6 HCP ELISA Kit, 2nd Generation, Semi-Quantitative Detection of HCP Impurities, Research and Manufacturing Use Only, 28 Units
Supplier:  Cygnus Technologies F1055
Encompass
The Cygnus Technologies PER.C6 HCP ELISA Kit, 2nd Generation is a sensitive immunoassay for the semi-quantitative detection of host cell protein impurities. Designed for products made using the PER.C6 human cell line, this kit enhances impurity monitoring during bioprocess development and product release. Its broad reactivity to PER.C6 HCPs, characterized by advanced techniques, makes it a versatile tool for both research and manufacturing applications, setting it apart from predecessors.
Catalog No. NC2682923