Learn More
TARGETMOL CHEMICALS INC Peramivir 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T2133910MG
Encompass_Preferred
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Peramivir (BCX1812) is a highly selective influenza A and B neuraminidase inhibitor with antiviral activity and anti-cytokine effects, which can be used in the study of novel coronavirus and influenza virus infections. Purity 99.82%
Catalog No. 50-287-7478