Learn More
Apexbio Technology LLC Peramivir(Synonyms: Rapivab, BCX-1812, RWJ-270201, Peramiflu), 10mM (in 1mL DMSO), CAS: 330600-85-6.
SDP
Supplier:  Apexbio Technology LLC A37095.1
Encompass_Preferred
Peramivir (CAS 330600-85-6) is a neuraminidase inhibitor under investigation for antiviral activity against influenza virus Structurally peramivir functions as a transition-state analogue inhibitor selectively targeting viral neuraminidase to obstruct enzymatic cleavage required for virion release from infected host cells By this action peramivir effectively impedes viral propagation It has undergone clinical evaluation in research contexts notably receiving emergency use authorization for hospitalized patients during the 2009 H1N1 influenza pandemic
Catalog No. 50-000-60302