Learn More
Medchemexpress LLC Peramivir | 330600-85-6 | 98.3% | 382.45 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY17015100MG
Encompass_Preferred
Peramivir trihydrate is a highly potent, selective, and orally active inhibitor of influenza virus neuraminidase, showing IC50 values from 0.9 to 4.3 nM across nine NA subtypes.
  • Highly potent, selective, and orally active influenza virus neuraminidase (NA) inhibitor.
  • Shows IC50 values ranging from 0.9 to 4.3 nM for nine NA subtypes.
  • Nontoxic to macrophages at concentrations up to 40 μM.
  • Inhibits cytokine release in LPS-induced hPBMCs.
  • Inhibits LPS-induced cytokine storm and attenuates acute lung injury in mice.
  • Prolongs survival in cytokine storm syndrome model mice.
  • Decreases various cytokines and chemokines, including TNF-a, IFN-a, and IL-6.
  • Provides significant protective effects on lung tissues.
  • Rescues BALB scid mice from lethal influenza B virus challenge in immunocompromised models.
Catalog No. 50-003-86053