Learn More
Medchemexpress LLC Peramivir trihydrate | 1041434-82-5 | 98.3% | 382.45 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1701510MM/1ML
Encompass_Preferred
Peramivir trihydrate (RWJ-270201 trihydrate) is a highly potent, selective, and orally active influenza virus neuraminidase (NA) inhibitor, with IC50 values ranging from 0.9 to 4.3 nM for nine NA subtypes. It is suitable for various research applications.
  • Targets influenza virus, IKK, JNK, STAT, p38 MAPK, ERK
  • Useful for infection and inflammation/immunology research
  • Cited in over 10 publications
Catalog No. 50-003-86054