Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions Perchloric Acid 0.010N in Acetic Acid, NIST Traceable (4L)
SDP

Supplier:  Aqua Solutions 66034L

Encompass_Preferred

Perchloric Acid 0.010N in Acetic Acid, NIST Traceable (4L)

Catalog No. NC9035540


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.