Learn More
Biolegend PerCP anti-human CD11a
Supplier:  Biolegend 350608
Encompass_Preferred
PerCP anti-human CD11a [TS2/4]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-168-618