Learn More
Biolegend PerCP anti-human CD45RO
Supplier:  Biolegend 304252
Encompass_Preferred
PerCP anti-human CD45RO [UCHL1]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Chimpanzee, Cynomolgus, Common Marmoset; Apps: FC; Size: 100 tests
Catalog No. 50-207-3080