Learn More
Biolegend PerCP anti-human CD45
Supplier:  Biolegend 368506
Encompass_Preferred
PerCP anti-human CD45 [2D1]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1298530