Learn More
Biolegend PerCP anti-mouse CD3ε
Supplier:  Biolegend 100325
Encompass_Preferred
PerCP anti-mouse CD3ε [145-2C11]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-162-400