Learn More
Cytek Biosciences Inc PerCP-Cy5.5 aMu MHCII Anti
Supplier: Cytek Biosciences Inc 655321U025
Product Name: PerCP-Cyanine5.5 Anti-Mouse MHC Class II (I-A/I-E) (M5/114.15.2)
Product Class: Antibody
Type: Primary
Product Name: PerCP-Cyanine5.5 Anti-Mouse MHC Class II (I-A/I-E) (M5/114.15.2)
Cytek Catalog Number: 65-5321-U025
Marker: MHC Class II (I-A/I-E)
Target Species: Mouse
Clonality: Monoclonal
Clone: M5/114.15.2
Host Species: Rat
Isotype: IgG2b, k
Conjugation: PerCP-Cyanine5.5
Size: 25 μg
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.