Learn More
Biolegend PerCP/Cyanine5.5 anti-human CD14
Supplier:  Biolegend 325622
Encompass_Preferred
PerCP/Cyanine5.5 anti-human CD14 [HCD14]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-167-305