Learn More
Biolegend PerCP/Cyanine5.5 anti-human CD34
Supplier:  Biolegend 343521
Encompass_Preferred
PerCP/Cyanine5.5 anti-human CD34 [581]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Cynomolgus; Apps: FC; Size: 25 tests
Catalog No. 50-168-200