Learn More
Biolegend PerCP/Cyanine5.5 anti-mouse CD68
Supplier:  Biolegend 137009
Encompass_Preferred
PerCP/Cyanine5.5 anti-mouse CD68 [FA-11]; Isotype: Rat IgG2a; Reactivity: Mouse; Apps: ICFC, FC; Size: 25 ug
Catalog No. 50-164-943