Learn More
Biolegend PerCP/Cyanine5.5 anti-MU/HU GL7 Antigen (T/B Cell Act. Marker)
Supplier:  Biolegend 144610
Encompass_Preferred
PerCP/Cyanine5.5 anti-MU/HU GL7 Antigen (T/B Cell Act. Marker) [GL7]; Isotype: Rat IgM, κ; Reactivity: Mouse, Human; Apps: FC; Size: 100 ug
Catalog No. 50-605-096