Learn More
Biolegend PerCP/Cyanine5.5 anti-human CD15 (SSEA-1)
Supplier:  Biolegend 323020
Encompass_Preferred
PerCP/Cyanine5.5 anti-human CD15 (SSEA-1) [W6D3]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-604-156