Learn More
Biolegend PerCP/FireTM 780 anti-human CD
Supplier:  Biolegend 393337
Encompass_Preferred
PerCP/Fire™ 780 anti-human CD57 Recombinant [QA17A04]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-279-6520