Learn More
Biolegend PerCP/FireTM 780 anti-mouse CD
Supplier:  Biolegend 156627
Encompass_Preferred
PerCP/Fire™ 780 anti-mouse CD16/32 [S17011E]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 25 μg
Catalog No. 50-279-6445