Learn More
Biolegend PerCP/FireTM 806 anti-human CD
Supplier:  Biolegend 350253
Encompass_Preferred
PerCP/Fire™ 806 anti-human CD103 (Integrin αE) [Ber-ACT8]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-290-6262