Learn More
ABclonal Technology PerCP Rabbit anti-Mouse CD8a
SDP
Supplier:  ABclonal Technology A27335100T
Encompass_Preferred
Enables identical protein binding activity Acts upstream of or within several processes including cytotoxic T cell differentiation defense response to virus and positive regulation of calcium-mediated signaling Located in external side of plasma membrane Is expressed in several structures including endocrine gland exocrine gland genitourinary system gut and hemolymphoid system gland Orthologous to human CD8A (CD8a molecule)
Catalog No. 50-000-09504