Learn More
Medchemexpress LLC HY-100008 5mg Medchemexpress, Peretinoin CAS:81485-25-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1000085MG
Encompass_Preferred
Medchemexpress, HY-100008 5mg Peretinoin CAS:81485-25-8 Peretinoin is an oral acyclic retinoid retinoid with a vitamin A-like structure that targets retinoid nuclear receptors such as retinoid X receptor (RXR) and retinoic acid receptor (RAR) . Peretinoin reduces the mRNA level of sphingosine kinase 1 (SPHK1) in vitro by downregulating a transcription factor, Sp1 [1] . Peretinoin prevents the progression of non-alcoholic steatohepatitis (NASH) and the development of hepatocellular carcinoma (HCC) through activating the autophagy pathway by increased Atg16L1 expression [2] . Peretinoin inhibits HCV RNA amplification and virus release by altering lipid metabolism with a EC 50 of 9 μM [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1685