Learn More
Medchemexpress LLC Perfluoro(2-methyl-3-oxahexanoic) acid | 13252-13-6 | MFCD00236734 | 99.4% | 330.05 g·mol⁻1 | C6HF11O3 | 25 G

Supplier: Medchemexpress LLC 25GHYW081067
Perfluoro(2-methyl-3-oxahexanoic) acid (HFPO-DA) is a perfluorinated carboxylic acid used as a reagent and as a raw material for producing fluorinated polymers, coatings, and surface treatments. It is persistent in the environment and should be handled according to the safety data sheet.
- High purity (99.4%).
- CAS number 13252-13-6.
- Molecular formula C6HF11O3 and molecular weight 330.05 g·mol⁻1.
- Supplied in laboratory pack sizes including 1 g, 5 g, 10 g, 25 g, 50 g, and 100 g.
- Certificate of analysis and safety data sheet available.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.