Learn More
Medchemexpress LLC Perfluorodecalin | 306-94-5 | MFCD00010626 | 98.0% | 462.08 | C10F18 | 5 G
SDP
Supplier:  Medchemexpress LLC HY1082985G
Encompass_Preferred
Perfluorodecalin is a perfluorocarbon liquid (C10F18) used as an oxygen-carrying solvent and research reagent. It dissolves large amounts of oxygen, is thermally stable and non-flammable, and is applied in biomedical research such as vitreoretinal surgery models and tissue oxygenation studies. Typical properties include CAS 306-94-5, molecular weight 462.08, and NMR purity ~98%.
  • High oxygen solubility for tissue and cell studies.
  • Thermally stable and non-flammable liquid.
  • Used in ophthalmic and vitreoretinal research models.
  • Functions as a fluorous-phase solvent for specialized chemistry.
  • High analytical purity for research applications.
  • Colorless to light yellow appearance.
Catalog No. 50-002-44003