Learn More
Strem, An Ascensus Company CAS# 355-42-0. 50g. Perfluorohexane, 98+% (85% n-isomer). MFCD00000437
SDP
Supplier:  Strem, An Ascensus Company 09607250G
Encompass
CAS# 355-42-0. 50g. Perfluorohexane, 98+% (85% n-isomer). MFCD00000437. Molecular Weight: 338.04. Molecular Formula: C6F14. Color/form: colorless liq. Strem# 09-6072. http://www.strem.com/catalog/v/09-6072/
Catalog No. 50-901-09395