Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Perfluorohexanesulfonamide | 41997-13-1 | MFCD09263736 | >75.0% | 399.13 g/mol | C6H2F13NO2S | 10 MG
SDP

Supplier:  Medchemexpress LLC HYW58780510MG

Encompass_Preferred

Perfluorohexanesulfonamide (FHxSA) is a perfluorinated sulfonamide used as a research reagent and reported inhibitor of carbonic anhydrase. It is supplied in small research-scale quantities for biochemical assays and PFAS-related studies, including enzymology and environmental fate investigations.

  • Used as a carbonic anhydrase inhibitor in biochemical assays.
  • Perfluorinated sulfonamide structure relevant to PFAS research.
  • Supplied in small, research-scale quantities appropriate for assay development.
  • Chemical identifiers: CAS 41997-13-1; formula C6H2F13NO2S.
  • Molecular weight approximately 399.13 g/mol.
  • Purity reported as >75% (depositor supplied); verify with supplier COA/SDS prior to use.

Catalog No. 50-002-84592


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.