Learn More
Cayman Chemical PerfluoronnoIc AcId 5g
SDP
Supplier:  Cayman Chemical 372505 G
Encompass_Preferred
A perfluoroalkyl substance; increases serum glucose and hepatic glycogen levels, as well as increases hepatic hydrogen peroxide and MDA levels, in rats at 5 mg/kg per day; induces spontaneous abortion in pregnant mice at 10 mg/kg per day; induces death of pups within 10 days at 5 mg/kg per day to dams on gestational days 1-17; has been found in bottled water, watersheds, and marine mammals
Catalog No. 50-291-9388