Learn More
TARGETMOL CHEMICALS INC Pergolide mesylate 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T222625MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg and bulk. Please contact Fisher for quotes. Pergolide mesylate salt(LY127809) is a long-acting dopamine agonist which has been used to treat PARKINSON DISEASE and HYPERPROLACTINEMIA but withdrawn from some markets due to potential for HEART VALVE DISEASES. Purity 99.09%
Catalog No. 50-287-9558