Learn More
Medchemexpress LLC Perillyl alcohol | 536-59-4 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN700010MM/1ML
Encompass_Preferred
Perillyl alcohol is an orally active monoterpene that exhibits various activities such as analgesic, anti-inflammatory, anti-tumor, anti-angiogenic, and anti-nociceptive effects. It can induce apoptosis and cell cycle arrest in tumor cells.
  • Increases caspase-3 activity and inhibits Akt phosphorylation in BLMVECs.
  • Reduces basal secretion of VEGF in various cell lines.
  • Induces apoptosis and suppresses Ras protein in melanoma cells.
  • Reduces blood vessels in the chicken embryo chorioallantoic membrane model.
  • Shows anti-melanoma activity and delays tumor development in TPras mouse model.
  • Possesses analgesic and anti-nociceptive activities in mice.
Catalog No. 50-004-19807