Learn More
Medchemexpress LLC Perinaphthenone | 548-39-0 | MFCD00004143 | 99.9% | 180.20 g/mol | C13H8O | 250 MG
SDP
Supplier:  Medchemexpress LLC HYW286805250MG
Encompass_Preferred
Perinaphthenone (phenalenone) is a small-molecule antifungal research reagent reported to inhibit growth of plant-pathogenic fungi. Reported IC50 values include 36.2 μM (day 3) for Botrytis spp., 13.3 μM (day 3) for Fusarium spp., and 39.0 μM (60 h) for Botryodiplodia spp. The compound is supplied as a dry solid and in DMSO solution for convenient reconstitution and assay use.
  • Exhibits antifungal activity against multiple fungal genera.
  • Provides clear inhibitory data for use in assay development.
  • Available as a solid and as a DMSO stock solution for convenience.
  • High reported purity suitable for research applications.
  • Useful for in vitro fungal growth inhibition and related bioassays.
Catalog No. 50-002-81879