Learn More
Medchemexpress LLC Perindopril-d4 | 1356929-58-2 | 99.3% | C19H28D4N2O5 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY143789S1MG
Encompass_Preferred
Perindopril-d4 is a deuterium-labeled version of Perindopril, an angiotensin-converting enzyme (ACE) inhibitor. Deuterium-labeled compounds are used in research as tracers for quantitation during drug development due to their potential to influence the pharmacokinetic and metabolic profiles of drugs. This stable isotope is suitable for research applications.
  • Stable isotope-labeled compound
  • Can be used as a tracer
  • Suitable as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-54551