Learn More
Apexbio Technology LLC Perindopril Erbumine 107133-36-8 100mg
SDP
Supplier:  Apexbio Technology LLC A8019100
Encompass_Preferred
Perindopril Erbumine (CAS 107133-36-8) is a potent inhibitor of angiotensin-converting enzyme (ACE) demonstrating an IC value of 1 05 nM This small molecule is brain penetrant and modulates the renin-angiotensin system within the central nervous system Experimental studies indicate that Perindopril Erbumine suppresses the elevation of hippocampal ACE activity and mitigates cognitive deficits in PS2APP-transgenic mouse models These properties support its utility in research investigating the interactions between ACE activity neurophysiology and cognitive function
Catalog No. 50-224-0729