Learn More
Medchemexpress LLC Perindopril erbumine | 107133-36-8 | 99.98% | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYB0130A
Encompass_Preferred
Perindopril erbumine is an angiotensin-converting enzyme inhibitor that modulates NF-κB and STAT3 signaling and inhibits glial activation and neuroinflammation. It is commonly utilized in the research of chronic kidney disease and high blood pressure.
  • Modulates NF-κB and STAT3 signaling
  • Inhibits glial activation and neuroinflammation
  • Suitable for chronic kidney disease research
  • Suitable for high blood pressure research
Catalog No. 50-004-05730