Learn More
Selleck Chemical LLC Perindopril Erbumine 50mg 107133-36-8 S9490-3
SDP
Supplier:  Selleck Chemical LLC S150650MG
Encompass_Preferred
Perindopril Erbumine (S9490-3) is a pro-drug and metabolized in vivo by hydrolysis of the ester group to form Perindoprilat, the biologically active¿metabolite, a potent ACE inhibitor with IC50 of 1.05 nM. Perindopril Erbumine is used for in vivo studies and Perindoprilat is recommened for in vitro research. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1332