Learn More
Genetex Inc Periostin Monoclonal Antibody [MSVA-649M] HistoMAX™ (GTX04460) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04460
Encompass_Preferred
The Periostin antibody [MSVA-649M] HistoMAX™ (GTX04460) is a Mouse Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human mucosa from rectum tissue using GTX04460 Periostin antibody [MSVA-649M] HistoMAX. A periostin positive subepithelial band is regularly seen in the colorectal mucosa. Additional periostin deposits occur in the lamina propria.
Catalog No. NC3667742